Assay Details
| Gene Symbol: RAMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098860.3 | NP_001092330.1 | 3 - 4 | 394 | 61 | |
| GenBank mRNA | BC146273.1 | - | 3 - 4 | 395 | 61 | |
| DT719941.1 | - | 3 - 4 | 372 | 61 | ||
| EE245593.1 | - | 3 - 4 | 315 | 61 | ||
More Information
Set Membership: |
|

RAMP2
Genomic Map