Assay Details
| Gene Symbol: GET3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098864.1 | NP_001092334.1 | 4 - 5 | 645 | 71 | |
| GenBank mRNA | BC142125.1 | - | 4 - 5 | 645 | 71 | |
More Information
Set Membership: |
|

GET3
Genomic Map