Assay Details
| Gene Symbol: ACP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098866.1 | NP_001092336.1 | 4 - 5 | 491 | 70 | |
| GenBank mRNA | BC146093.1 | - | 4 - 5 | 491 | 70 | |
More Information
Set Membership: |
|

ACP3
Genomic Map