Assay Details
| Gene Symbol: NARS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098867.2 | NP_001092337.1 | 13 - 14 | 1577 | 70 | |
| GenBank mRNA | BC142045.1 | - | 13 - 14 | 1578 | 70 | |
More Information
Set Membership: |
|

NARS2
Genomic Map