Assay Details
| Gene Symbol: ZFAND5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101045.1 | NP_001094515.1 | 2 - 3 | 202 | 70 | |
| GenBank mRNA | BC151457.1 | - | 2 - 3 | 202 | 70 | |
More Information
Set Membership: |
|

ZFAND5
Genomic Map