Assay Details
| Gene Symbol: PDHA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101046.2 | NP_001094516.1 | 10 - 11 | 1107 | 83 | |
| GenBank mRNA | BC114120.1 | - | 10 - 11 | 1108 | 83 | |
| BC151313.1 | - | 10 - 11 | 1109 | 83 | ||
More Information
Set Membership: |
|

PDHA1
Genomic Map