Assay Details
| Gene Symbol: UPB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101050.1 | NP_001094520.1 | 4 - 5 | 475 | 54 | |
| GenBank mRNA | BC151517.1 | - | 4 - 5 | 475 | 54 | |
More Information
Set Membership: |
|

UPB1
Genomic Map