Assay Details
| Gene Symbol: HSD17B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102365.1 | NP_001095835.1 | 3 - 4 | 1021 | 56 | |
| GenBank mRNA | AF265564.1 | - | 3 - 4 | 420 | 56 | |
| BC116044.1 | - | 3 - 4 | 1021 | 56 | ||
More Information
Set Membership: |
|

HSD17B1
Genomic Map