Assay Details
| Gene Symbol: ST8SIA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103227.1 | NP_001096697.1 | 6 - 7 | 1015 | 95 | |
| GenBank mRNA | AJ868431.1 | - | 6 - 7 | 632 | 95 | |
| BC140641.1 | - | 6 - 7 | 1015 | 95 | ||
More Information
Set Membership: |
|

ST8SIA6
Genomic Map