Assay Details
| Gene Symbol: LTBR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103228.2 | NP_001096698.1 | 8 - 9 | 983 | 65 | |
| GenBank mRNA | BC149639.1 | - | 8 - 9 | 984 | 65 | |
More Information
Set Membership: |
|

LTBR
Genomic Map