Assay Details
| Gene Symbol: KRT17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105322.1 | NP_001098792.1 | 7 - 8 | 1267 | 86 | |
| GenBank mRNA | BC142137.1 | - | 7 - 8 | 1267 | 86 | |
| BT029882.1 | - | 7 - 8 | 1242 | 86 | ||
More Information
Set Membership: |
|

KRT17
Genomic Map