Assay Details
| Gene Symbol: IGFBP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105327.2 | NP_001098797.1 | 3 - 4 | 718 | 76 | |
| GenBank mRNA | BC149394.1 | - | 3 - 4 | 720 | 76 | |
More Information
Set Membership: |
|

IGFBP5
Genomic Map