Assay Details
| Gene Symbol: CENPP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105615.1 | NP_001099085.1 | 7 - 8 | 756 | 80 | |
| GenBank mRNA | BC134699.1 | - | 7 - 8 | 756 | 80 | |
More Information
Set Membership: |
|

CENPP
Genomic Map