Assay Details
| Gene Symbol: MCAM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110062.1 | NP_001103532.1 | 11 - 12 | 2063 | 99 | |
| GenBank mRNA | BC118105.1 | - | 11 - 12 | 2063 | 99 | |
More Information
Set Membership: |
|

MCAM
Genomic Map