Assay Details
| Gene Symbol: TPPP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173976.3 | NP_776401.1 | 1 - 2 | 308 | 73 | |
| GenBank mRNA | BC133343.1 | - | 2 - 3 | 662 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TPPP| Gene Symbol: TPPP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173976.3 | NP_776401.1 | 1 - 2 | 308 | 73 | |
| GenBank mRNA | BC133343.1 | - | 2 - 3 | 662 | 73 | |
Set Membership: |
|