Assay Details
| Gene Symbol: CTSB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174031.2 | NP_776456.1 | 2 - 3 | 212 | 65 | |
| GenBank mRNA | BC102997.1 | - | 3 - 4 | 274 | 65 | |
| L06075.1 | - | 2 - 3 | 212 | 65 | ||
More Information
Set Membership: |
|

CTSB
Genomic Map