Assay Details
| Gene Symbol: FGF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174056.4 | NP_776481.2 | 2 - 3 | 378 | 137 | |
| GenBank mRNA | KX834563.1 | - | 2 - 3 | 275 | 137 | |
| M13440.1 | - | 2 - 3 | 378 | 137 | ||
More Information
Set Membership: |
|

FGF2
Genomic Map