Assay Details
| Gene Symbol: BFSP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174248.3 | NP_776673.1 | 5 - 6 | 1032 | 62 | |
| GenBank mRNA | BC122762.1 | - | 5 - 6 | 1032 | 62 | |
| X75160.1 | - | 1079 | 62 | |||
More Information
Set Membership: |
|

BFSP2
Genomic Map