Assay Details
| Gene Symbol: CXCL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174300.2 | NP_776725.1 | 1 - 2 | 158 | 150 | |
| GenBank mRNA | AF149249.1 | - | 1 - 2 | 158 | 150 | |
| BC142108.1 | - | 1 - 2 | 182 | 150 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC142108.1, NM_174300.2 transcripts.
Set Membership: |
|

CXCL5
Genomic Map