Assay Details
| Gene Symbol: EDNRA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174308.2 | NP_776733.1 | 6 - 7 | 1488 | 73 | |
| GenBank mRNA | BC133407.1 | - | 5 - 6 | 1037 | 73 | |
| BC142309.1 | - | 6 - 7 | 1502 | 73 | ||
| X57765.1 | - | 1502 | 73 | |||
More Information
Set Membership: |
|

EDNRA
Genomic Map