Assay Details
| Gene Symbol: IKBKG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174354.3 | NP_776779.1 | 4 - 5 | 587 | 60 | |
| GenBank mRNA | AJ414557.1 | - | 3 - 4 | 518 | 60 | |
| BC133306.1 | - | 4 - 5 | 587 | 60 | ||
| BT020753.1 | - | 3 - 4 | 524 | 60 | ||
| BT020767.1 | - | 3 - 4 | 462 | 60 | ||
| BT020840.1 | - | 3 - 4 | 517 | 60 | ||
| BT020872.1 | - | 3 - 4 | 470 | 60 | ||
More Information
Set Membership: |
|

IKBKG
Genomic Map