Assay Details
| Gene Symbol: IRAG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174392.4 | NP_776817.2 | 9 - 10 | 1650 | 73 | |
| NM_194465.4 | NP_919447.2 | 9 - 10 | 1360 | 73 | ||
| GenBank mRNA | AF195526.1 | - | 9 - 10 | 1650 | 73 | |
| AF195527.1 | - | 9 - 10 | 1360 | 73 | ||
| BC153850.1 | - | 1 - 2 | 463 | 73 | ||
More Information
Set Membership: |
|

IRAG1
Genomic Map