Assay Details
| Gene Symbol: SELENOP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174459.3 | NP_776884.2 | 1 - 2 | 156 | 69 | |
| GenBank mRNA | BC102267.1 | - | 1 - 2 | 54 | 69 | |
| CB532844.1 | - | 1 - 2 | 156 | 69 | ||
| EE223056.1 | - | 1 - 2 | 54 | 69 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_174459.3 transcripts.
Set Membership: |
|

SELENOP
Genomic Map