Assay Details
| Gene Symbol: SELENOP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174459.3 | NP_776884.2 | 3 - 4 | 590 | 119 | |
| GenBank mRNA | AB032826.1 | - | 2 - 3 | 418 | 119 | |
| CB532844.1 | - | 3 - 4 | 590 | 119 | ||
| D25220.1 | - | 3 - 4 | 461 | 119 | ||
| EE223056.1 | - | 3 - 4 | 488 | 119 | ||
More Information
Set Membership: |
|

SELENOP
Genomic Map