Assay Details
| Gene Symbol: SFTPC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174462.4 | NP_776887.1 | 5 - 6 | 607 | 83 | |
| GenBank mRNA | AF250035.1 | - | 5 - 6 | 582 | 83 | |
| BC123536.1 | - | 5 - 6 | 608 | 83 | ||
More Information
Set Membership: |
|

SFTPC
Genomic Map