Assay Details
| Gene Symbol: DNAJC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174533.4 | NP_776958.2 | 2 - 3 | 341 | 68 | |
| NR_120605.1 | 1 - 2 | 121 | 68 | |||
| GenBank mRNA | BC120234.1 | - | 2 - 3 | 332 | 68 | |
| EE238321.1 | - | 2 - 3 | 342 | 68 | ||
| X92666.1 | - | 110 | 68 | |||
| X92667.1 | - | 110 | 68 | |||
More Information
Set Membership: |
|

DNAJC5
Genomic Map