Assay Details
| Gene Symbol: IGFBP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174557.4 | NP_776982.1 | 2 - 3 | 749 | 60 | |
| GenBank mRNA | BC140688.1 | - | 2 - 3 | 750 | 60 | |
| S52770.1 | - | 2 - 3 | 713 | 60 | ||
More Information
Set Membership: |
|

IGFBP4
Genomic Map