Assay Details
| Gene Symbol: SLC2A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174603.3 | NP_777028.2 | 7 - 8 | 1076 | 62 | |
| GenBank mRNA | AY033938.1 | - | 7 - 8 | 1042 | 62 | |
| BC133291.1 | - | 7 - 8 | 1076 | 62 | ||
More Information
Set Membership: |
|

SLC2A3
Genomic Map