Assay Details
| Gene Symbol: FST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175801.3 | NP_786995.2 | 3 - 4 | 684 | 60 | |
| GenBank mRNA | BC133637.1 | - | 3 - 4 | 685 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: FST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175801.3 | NP_786995.2 | 3 - 4 | 684 | 60 | |
| GenBank mRNA | BC133637.1 | - | 3 - 4 | 685 | 60 | |
Set Membership: |
|