Assay Details
| Gene Symbol: BANF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182988.3 | NP_892033.1 | 1 - 2 | 115 | 64 | |
| GenBank mRNA | BT020975.1 | - | 1 - 2 | 58 | 64 | |
| DN529481.1 | - | 1 - 2 | 51 | 64 | ||
| DN639680.1 | - | 1 - 2 | 115 | 64 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_182988.3 transcripts.
Set Membership: |
|

BANF1
Genomic Map