Assay Details
| Gene Symbol: ZNF692 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024496.2 | NP_001019667.2 | 1 - 2 | 52 | 56 | |
| GenBank mRNA | BC126714.1 | - | 1 - 2 | 52 | 56 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC126714.1, NM_001024496.2 transcripts.
Set Membership: |
|

ZNF692
Genomic Map