Assay Details
| Gene Symbol: ZNF692 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024496.2 | NP_001019667.2 | 8 - 9 | 1006 | 75 | |
| GenBank mRNA | BC126714.1 | - | 8 - 9 | 1006 | 75 | |
| BT021686.1 | - | 8 - 9 | 1028 | 75 | ||
More Information
Set Membership: |
|

ZNF692
Genomic Map