Assay Details
| Gene Symbol: DPP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024513.2 | NP_001019684.2 | 12 - 13 | 1436 | 59 | |
| GenBank mRNA | AW654853.1 | - | 2 - 3 | 185 | 59 | |
| BC102861.1 | - | 12 - 13 | 1427 | 59 | ||
More Information
Set Membership: |
|

DPP3
Genomic Map