Assay Details
| Gene Symbol: FKBP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034322.2 | NP_001029494.1 | 9 - 10 | 1320 | 71 | |
| GenBank mRNA | BC102456.1 | - | 9 - 10 | 1336 | 71 | |
| BT030531.1 | - | 9 - 10 | 1320 | 71 | ||
More Information
Set Membership: |
|

FKBP4
Genomic Map