Assay Details
| Gene Symbol: KLF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035271.4 | NP_001030348.2 | 1 - 2 | 311 | 114 | |
| GenBank mRNA | BC107527.1 | - | 1 - 2 | 313 | 114 | |
More Information
Set Membership: |
|

KLF6
Genomic Map