Assay Details
Gene Symbol: RAD18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035295.3 | NP_001030372.2 | 7 - 8 | 947 | 85 | |
GenBank mRNA | BC109568.1 | - | 7 - 8 | 948 | 85 | |
BT021682.1 | - | 7 - 8 | 899 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
