Assay Details
| Gene Symbol: TBATA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038121.2 | NP_001033210.2 | 6 - 7 | 821 | 63 | |
| GenBank mRNA | BC110031.1 | - | 6 - 7 | 822 | 63 | |
| CK774174.1 | - | 3 - 4 | 425 | 63 | ||
More Information
Set Membership: |
|

TBATA
Genomic Map