Assay Details
| Gene Symbol: VPS26C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040510.2 | NP_001035600.2 | 1 - 2 | 95 | 98 | |
| GenBank mRNA | BT021567.1 | - | 1 - 2 | 95 | 98 | |
More Information
Set Membership: |
|

VPS26C
Genomic Map