Assay Details
| Gene Symbol: VPS26C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040510.2 | NP_001035600.2 | 3 - 4 | 385 | 58 | |
| GenBank mRNA | BC148109.1 | - | 3 - 4 | 397 | 58 | |
| BF074212.1 | - | 1 - 2 | 38 | 58 | ||
| BT021567.1 | - | 3 - 4 | 385 | 58 | ||
More Information
Set Membership: |
|

VPS26C
Genomic Map