Assay Details
| Gene Symbol: VPS26C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040510.2 | NP_001035600.2 | 4 - 5 | 464 | 60 | |
| GenBank mRNA | BC148109.1 | - | 4 - 5 | 476 | 60 | |
| BF074212.1 | - | 2 - 3 | 117 | 60 | ||
| BT021567.1 | - | 4 - 5 | 464 | 60 | ||
More Information
Set Membership: |
|

VPS26C
Genomic Map