Assay Details
| Gene Symbol: VPS26C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040510.2 | NP_001035600.2 | 6 - 7 | 696 | 104 | |
| GenBank mRNA | BC148109.1 | - | 6 - 7 | 708 | 104 | |
| BF074212.1 | - | 4 - 5 | 349 | 104 | ||
| BT021567.1 | - | 6 - 7 | 696 | 104 | ||
More Information
Set Membership: |
|

VPS26C
Genomic Map