Assay Details
| Gene Symbol: VPS26C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040510.2 | NP_001035600.2 | 7 - 8 | 846 | 92 | |
| GenBank mRNA | BC148109.1 | - | 7 - 8 | 858 | 92 | |
| BT021567.1 | - | 7 - 8 | 845 | 92 | ||
More Information
Set Membership: |
|

VPS26C
Genomic Map