Assay Details
| Gene Symbol: GPATCH11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046127.3 | NP_001039592.3 | 6 - 7 | 639 | 62 | |
| GenBank mRNA | BC112802.1 | - | 6 - 7 | 640 | 62 | |
More Information
Set Membership: |
|

GPATCH11
Genomic Map