Assay Details
| Gene Symbol: C5H22orf23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046253.3 | NP_001039718.2 | 3 - 4 | 377 | 79 | |
| NM_001314055.1 | NP_001300984.1 | 3 - 4 | 356 | 79 | ||
| NR_132765.1 | 3 - 4 | 377 | 79 | |||
| GenBank mRNA | BC110234.1 | - | 3 - 4 | 297 | 79 | |
| DT812245.1 | - | 3 - 4 | 344 | 79 | ||
| EH138794.1 | - | 2 - 3 | 266 | 79 | ||
| EH141917.1 | - | 3 - 4 | 339 | 79 | ||
More Information
Set Membership: |
|

C5H22orf23
Genomic Map