Assay Details
| Gene Symbol: C5H22orf23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046253.3 | NP_001039718.2 | 6 - 7 | 795 | 66 | |
| NM_001314055.1 | NP_001300984.1 | 6 - 7 | 774 | 66 | ||
| NR_132765.1 | 6 - 7 | 880 | 66 | |||
| GenBank mRNA | BC110234.1 | - | 6 - 7 | 800 | 66 | |
| DT812245.1 | - | 6 - 7 | 762 | 66 | ||
| EH141917.1 | - | 6 - 7 | 757 | 66 | ||
More Information
Set Membership: |
|

C5H22orf23
Genomic Map