Assay Details
| Gene Symbol: TARBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075678.2 | NP_001069146.2 | 7 - 8 | 851 | 66 | |
| GenBank mRNA | BC122653.1 | - | 7 - 8 | 851 | 66 | |
More Information
Set Membership: |
|

TARBP2
Genomic Map