Assay Details
| Gene Symbol: CA5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080908.3 | NP_001074377.2 | 1 - 2 | 70 | 61 | |
| GenBank mRNA | BT026329.1 | - | 1 - 2 | 70 | 61 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001080908.3 transcripts.
Set Membership: |
|

CA5B
Genomic Map