Assay Details
| Gene Symbol: CA5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080908.3 | NP_001074377.2 | 4 - 5 | 590 | 72 | |
| GenBank mRNA | DV876521.1 | - | 1 - 2 | 113 | 72 | |
More Information
Set Membership: |
|

CA5B
Genomic Map