Assay Details
| Gene Symbol: CA5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080908.3 | NP_001074377.2 | 6 - 7 | 744 | 92 | |
| GenBank mRNA | DV876521.1 | - | 3 - 4 | 267 | 92 | |
More Information
Set Membership: |
|

CA5B
Genomic Map