Assay Details
| Gene Symbol: CA5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080908.3 | NP_001074377.2 | 7 - 8 | 904 | 63 | |
| GenBank mRNA | DV876521.1 | - | 4 - 5 | 427 | 63 | |
More Information
Set Membership: |
|

CA5B
Genomic Map